Learn More
Medchemexpress LLC Enzalutamide carboxylic acid | 1242137-15-0 | MFCD28411442 | 98.1% | 451.39 | C20H13F4N3O3S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY70002B10MG
Encompass_Preferred
Enzalutamide carboxylic acid is the inactive carboxylic-acid metabolite of the androgen receptor antagonist enzalutamide. It is supplied as a reference standard and research reagent for drug metabolism, pharmacology, and analytical method development.
  • Reference standard for metabolite identification and quantification.
  • High purity suitable for analytical and research applications.
  • Characterized by CAS 1242137-15-0 and formula C20H13F4N3O3S.
  • Provided in small, sealed quantities for laboratory use (10 MG typical).
Catalog No. 50-002-97930