Learn More
Medchemexpress LLC Enzalutamide carboxylic acid-d6 | 1242137-15-0 | 98.99% | C20H7D6F4N3O3S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY70002BS1MG
Encompass_Preferred
Enzalutamide carboxylic acid-d6 is the deuterium-labeled Enzalutamide carboxylic acid (MDV3100 carboxylic acid). Enzalutamide carboxylic acid is an inactive metabolite of Enzalutamide. This product is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-33821