Learn More
Medchemexpress LLC Enzalutamide (standard) | 915087-33-1 | 100.0% | C21H16F4N4O2S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY70002R
Encompass_Preferred
Enzalutamide (Standard) is the analytical standard of Enzalutamide, intended for research and analytical applications. It acts as an androgen receptor (AR) antagonist with an IC50 of 36 nM in LNCaP prostate cells and is also an autophagy activator. This analytical standard grade compound serves as a reference for assays, commonly utilized in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
  • Analytical standard of enzalutamide
  • Intended for research and analytical applications
  • Functions as an androgen receptor (AR) antagonist
  • Autophagy activator
  • Serves as a reference standard for assays
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Compatible with techniques such as HPLC, GC, and MS
Catalog No. 50-003-59258