Learn More
Medchemexpress LLC Enzalutamide (standard) | 915087-33-1 | 100.0% | C21H16F4N4O2S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY70002R50MG
Encompass_Preferred
Enzalutamide (Standard) is an analytical standard for Enzalutamide, also known as MDV3100. It is an androgen receptor (AR) antagonist with an IC50 of 36 nM in LNCaP prostate cells and an autophagy activator. This compound is designed for research and analytical applications.
  • Analytical standard of Enzalutamide
  • Androgen receptor (AR) antagonist
  • Autophagy activator
  • Reference for qualitative, quantitative, and methodological research (HPLC, GC, MS)
Catalog No. 50-004-33880