Learn More
Medchemexpress LLC Enzalutamide (Standard) | 915087-33-1 | 99.98% | C21H16F4N4O2S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY70002R100MG
Encompass_Preferred
Enzalutamide (Standard) is the analytical standard of Enzalutamide, intended for research and analytical applications. It functions as an androgen receptor (AR) antagonist with an IC50 of 36 nM in LNCaP prostate cells and is also an autophagy activator. The compound serves as a reference standard supplied for assays and is commonly utilized in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
  • For research and analytical applications
  • Androgen receptor (AR) antagonist
  • Autophagy activator
  • Analytical standard
  • Reference standard for assays
  • Utilized in qualitative, quantitative, and methodological research experiments
Catalog No. 50-004-34295