Learn More
Medchemexpress LLC Enzastaurin | 170364-57-5 | MFCD11040980 | >98.0% | 515.61 | C32H29N5O2 | 200 MG

Supplier: Medchemexpress LLC HY10342200MG
Enzastaurin is a small-molecule inhibitor selective for protein kinase C beta (PKCβ), used in biochemical and cellular research to probe PKC-related signaling, autophagy, and apoptosis. It exhibits a reported IC50 of ~6 nM against PKCβ and reduced potency against PKCα, PKCγ, and PKCε.
- Potent PKCβ inhibitor (IC50 ~6 nM).
- Useful for studies of autophagy, apoptosis, and cancer signaling.
- Chemical formula C32H29N5O2; molecular weight 515.61 g/mol.
- CAS number 170364-57-5.
- Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (1 year) or -20°C (6 months).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.