Learn More
Medchemexpress LLC Enzastaurin | 170364-57-5 | MFCD11040980 | >98.0% | 515.61 g/mol | C32H29N5O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10342100MG
Encompass_Preferred
Enzastaurin (LY317615) is a small-molecule research reagent that selectively inhibits protein kinase C beta (PKCβ). It is supplied as a powder for laboratory use and is commonly used in kinase inhibition and cell-signaling studies.
  • Potent PKCβ inhibitor with IC50 = 6 nM.
  • Shows 6- to 20-fold selectivity over PKCα, PKCγ, and PKCε.
  • Molecular formula C32H29N5O2 and molecular weight 515.61 g/mol.
  • Provided as a powder suitable for preparing stock solutions.
  • Used for kinase signaling, apoptosis, and autophagy research applications.
Catalog No. 50-002-44674