Learn More
Selleck Chemical LLC Enzastaurin S1055-1g
SDP
Supplier:  Selleck Chemical LLC S10551G
Encompass_Preferred
Enzastaurin is a potent PKC selective inhibitor with IC50 of 6 nM in cell-free assays 6- to 20-fold selectivity against PKC PKC and PKC Phase 3
Catalog No. 50-310-8521