Learn More
Medchemexpress LLC Enzomenib | 2412555-70-3 | 99.9% | 590.73 | C33H43FN6O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15679410MG
Encompass_Preferred
Enzomenib (DSP-5336) is a selective small-molecule inhibitor of the menin-MLL interaction used in research on hematological malignancies. Provided as a high-purity solid for in vitro and preclinical studies.
  • High purity (99.9%).
  • Supplied as a 10 MG quantity suitable for small-scale studies.
  • Chemical formula: C33H43FN6O3; molecular weight: 590.73.
  • Suitable for biochemical, cellular, and preclinical assays.
  • Safety and identification data available in the SDS and product literature.
Catalog No. 50-002-88215