Learn More
Bioassay Systems EnzyChrom™ L-Carnitine Assay Kit, For determination of L-Carnitine in urine, serum, plasma, cell lysates, tissue lysates and food products (e.g. milk).
Supplier: Bioassay Systems ELCR100
L-CARNITINE is found in most mammals, plants, and some bacteria. L-Carnitine’s most crucial role is in the transport of long-chain fatty acids from the cytosol into the mitochondria for fatty acid oxidation. BioAssay Systems’ method provides a simple and high-throughput assay for measuring L-Carnitine. In this assay, a multistep reaction produces H2O2 which reacts with a specific dye to form a pink colored product. The optical density at 570nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to the L-Carnitine concentration in the sample.