Learn More
Waters Corp WATERS CORPORATION Enzymate Pepsin Column, 300 Å, 5 μm, 2.1 mm x 30 mm
Supplier: Waters Corp 186007233
The Enzymate column is designed for the on-column digestion of proteins into peptides in an online format. It can be connected directly to an HDX system to feed digested peptides for characterizing sample proteins. The immobilized pepsin offers high enzymatic activity, allowing proteins to be efficiently digested as they pass through, streamlining the workflow by combining digestion and separation steps and facilitating downstream peptide mapping and protein sequencing applications.
- Designed for online digestion of intact proteins into peptides.
- Chemistry: Pepsin.
- Separation mode: Protein digestion.
- Particle substrate: Silica.
- pH range: 1 to 8.
- Particle shape: Spherical.
- Endfitting type: Direct-Connect.
- QC tested: Protein.
- Surface area: 185.
- System: HPLC.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.