Learn More
SEKISUI DIAGNOSTICS LLC Enzymatic Creatinine R1, 3 x 100 mL
Supplier:  SEKISUI DIAGNOSTICS LLC 26530
Encompass_Preferred
Sekisui’s Enzymatic Creatinine assay utilizes a multi-step approach ending with a photometric end-point reaction. The assay accurately measures creatinine levels in serum, plasma and urine.
Catalog No. NC0938883