Learn More
Eurofins Discoverx DYRK1A Kinase Enzyme Activity Assay Kit
Supplier:  Eurofins Discoverx 44951
Encompass
The DYRK1A Kinase Enzyme Activity Assay Kit provides a fully validated, ready-to-use biochemical assay built on the trusted ADP Hunter™ Plus platform from DiscoverX, enabling accurate assessment of kinase inhibitor potency. This comprehensive kit includes recombinant protein, substrate, ATP, detection reagents, and assay plates for precise measurement of ADP formation.
Catalog No. 50-004-56081