Learn More
Eurofins Discoverx Fyn Kinase Enzyme Activity Assay Kit
Supplier: Eurofins Discoverx 44441
The Fyn Kinase Enzyme Activity Assay Kit provides a fully validated, ready-to-use biochemical assay built on the trusted ADP Hunter™ Plus platform from DiscoverX, enabling accurate assessment of kinase inhibitor potency. This comprehensive kit includes recombinant protein, substrate, ATP, detection reagents, and assay plates for precise measurement of ADP formation.