Learn More
Eurofins Discoverx CK1e Kinase Enzyme Activity Assay Kit
Supplier:  Eurofins Discoverx 46045
Encompass
The CK1e Kinase Enzyme Activity Assay Kit provides a fully validated, ready-to-use biochemical assay built on the trusted ADP Hunter™ Plus platform from DiscoverX, enabling accurate assessment of kinase inhibitor potency. This comprehensive kit includes recombinant protein, substrate, ATP, detection reagents, and assay plates for precise measurement of ADP formation.
Catalog No. 50-004-55581