Learn More
Cusabio Technology LLC Human eosinophil cationic protein,ECP ELISA Kit
Supplier:  Cusabio Technology LLC CSBE11729H
Encompass_Preferred
This Human ECP ELISA Kit was designed for the quantitative measurement of Human ECP protein in serum, plasma, tissue homogenates. It is a Sandwich ELISA kit, its detection range is 1.56 ng/mL-100 ng/mL and the sensitivity is 0.39 ng/mL. One kit contains 96 wells.
Catalog No. NC2952935