Learn More
Scytek Laboratories Eosinophil - Mast Cell Stain Kit: Large
SDP
Supplier:  Scytek Laboratories CEM1
Encompass_Preferred
The Eosinophil - Mast Cell Stain Kit is intended for simultaneous visualization of eosinophils and mast cells.Mast Cells: Bright BlueEosinophils: Bright RedNuclei: BlueKit contents: AST125, Astra Blue Solution, 125 ml. VRN125, Vital New Red Solution, 125 ml. HMM125, Hematoxylin, Mayer’s (Lillie’s Mod.), 125 ml.
Catalog No. NC1969841