Learn More
Scytek Laboratories Eosinophil - Mast Cell Stain Kit: Small
SDP
Supplier:  Scytek Laboratories CEM2
Encompass_Preferred
The Eosinophil - Mast Cell Stain Kit is intended for simultaneous visualization of eosinophils and mast cells.Mast Cells: Bright BlueEosinophils: Bright RedNuclei: BlueKit contents: AST030, Astra Blue Solution, 30 ml.VRN030, Vital New Red Solution, 30 ml.HMM030, Hematoxylin, Mayer’s (Lillie’s Mod.), 30 ml.
Catalog No. NC1969842