Learn More
Cayman Chemical EP4 Receptor (C-Term) Blocking Peptide, 1 ea
SDP
Catalog No. 502988221
Encompass_Preferred
Peptide Sequence: human EP4 receptor sequence amino acids 459-488 (GSGRAGPAPKGSSLQVTFPSETLNLSEKCI) · To be used in conjunction with Cayman’s EP4 Receptor Polyclonal Antibody (Item No. 101775) to block protein-antibody complex formation during immunochemical analysis for the EP4 receptor.
Catalog No. 50-298-8221 Supplier Cayman Chemical Supplier No. 3017751 EA
Peptide Sequence: human EP4 receptor sequence amino acids 459-488 (GSGRAGPAPKGSSLQVTFPSETLNLSEKCI) · To be used in conjunction with Cayman’s EP4 Receptor Polyclonal Antibody (Item No. 101775) to block protein-antibody complex formation during immunochemical analysis for the EP4 receptor.