Learn More
Crescent Chemical Co Inc EPA 506 PHTH QC MIX 7COMP 1ML
SDP
Supplier:  Crescent Chemical Co Inc CTCC2027
Encompass_Preferred
EPA Method 506 - Phthalate QC Mix, 7 Components Benzyl butyl phthalate-250 ugmL, Bis(2-ethylhexyl)adipate-1,200 ugmL, Bis(2-ethylhexyl)phthalate-250 ugmL, Di-n-butyl phthalate-100 ugmL, Diethyl phthalate-100 ugmL, Dimethyl phthalate-100 ugmL, Di-n-oc
Catalog No. 50-678-24