Learn More
Crescent Chemical Co Inc EPA 8270 TUNING 50PPM 4COMP1ML
SDP
Supplier:  Crescent Chemical Co Inc CTCC2140
Encompass_Preferred
CLP Semivolatiles Complete Priority Pollutants Kits - GCMS Tuning Standard, 4 components Benzidine, DFTPP, p,p CAS Size - 1mL Catalog Number - CC2140
Catalog No. 50-680-26