Learn More
Selleck Chemical LLC Epacadostat
SDP
Supplier:  Selleck Chemical LLC S791025MG
Encompass_Preferred
Epacadostat (INCB024360) is a potent and selective indoleamine 2 3-dioxygenase (IDO1) inhibitor with IC50 of 10 nM and displays high selectivity over other related enzymes such as IDO2 or tryptophan 2 3-dioxygenase (TDO)
Catalog No. 50-000-03414