Learn More
ABclonal Technology EPB41 Rabbit mAb
SDP
Supplier:  ABclonal Technology A26853100UL
Encompass_Preferred
The protein encoded by this gene together with spectrin and actin constitute the red cell membrane cytoskeletal network This complex plays a critical role in erythrocyte shape and deformability Mutations in this gene are associated with type 1 elliptocytosis (EL1) Alternatively spliced transcript variants encoding different isoforms have been described for this gene
Catalog No. 50-000-08417