Learn More
Genetex Inc EpCAM Monoclonal Antibody [MSVA-326R] HistoMAX™ (GTX04380) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04380
Encompass_Preferred
The EpCAM antibody [MSVA-326R] HistoMAX™ (GTX04380) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human epithelioid malignant mesothelioma tissue using GTX04380 EpCAM antibody [MSVA-326R] HistoMAX. EpCAM immunostaining is lacking in an epitheloid malignant mesothelioma.
Catalog No. NC3666291