Learn More
Genscript Corporation EpCAM/TROP1Biotin His Avi
Supplier:  Genscript Corporation Z04446100
Encompass
Epithelial Cellular Adhesion Molecule (EpCAM) also known as KS1/4 gp40 GA733-2 17-1A and TROP 1 is a 40 kDa transmembrane glycoprotein that consists of a 242 amino acid (aa) extracellular domain with two EGF like repeats a 23 aa transmembrane segment and a 26 aa cytoplasmic domain
Catalog No. 50-000-58481