Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Sanitary Gasket, EPDM, 2"" Tri-Clamp; 10/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3054866/DEL
Encompass
These gaskets are for Tri-Clamp style fittings and come in a wide variety of materials to match your specific process needs. All gaskets meet USDA, 3-A and CGMP criteria. FDA Compliant. All silicone, EPDM, PTFE, and Viton gaskets are animal-derived ingredient free and meet USP Class IV and cytotoxicity criteria. Contact for other sizes or materials. Platinum-cured silicone is ideal for most pharmaceutical, food, and beverage processing applications. Odorless and tasteless to ensure media purity. May undergo SIP. Usable from -58 to 446¿F (-50 to 230¿C). Viton offers excellent chemical resistance, especially to many acids and alkalis.
Catalog No. NC1579398
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.