Learn More
Medchemexpress LLC Eperisone hydrochloride | 56839-43-1 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB190110MM/1ML
Encompass_Preferred
Eperisone hydrochloride is an orally active antispastic agent with a vasodilator effect, commonly used in research for muscle stiffness and pain. It acts as a potent and selective P2X7 receptor antagonist and also demonstrates antagonism for human P2X3.
  • Relaxes skeletal and vascular smooth muscles
  • Reduces myotonia
  • Improves circulation
  • Suppresses pain reflex
  • Suppresses lung fibroblast activity
Catalog No. 50-004-11824