Learn More
Medchemexpress LLC HY-107367A 5mg Medchemexpress, Epertinib (hydrochloride) CAS:2071195-74-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107367A5MG
Encompass_Preferred
Medchemexpress, HY-107367A 5mg Epertinib (hydrochloride) CAS:2071195-74-7 Epertinib hydrochloride is a potent, orally active, reversible, and selective tyrosine kinase inhibitor of EGFR , HER2 and HER4 , with IC 50 s of 1.48 nM, 7.15 nM and 2.49 nM, respectively. Epertinib shows potent antitumor activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2283