Learn More
Medchemexpress LLC Epetraborole (hydrochloride) | 1234563-16-6 | MFCD30343855 | 99.6% | 273.52 | C11H17BClNO4 | 5MG
SDP
Supplier:  Medchemexpress LLC HY12479A5MG
Encompass_Preferred
Epetraborole (hydrochloride) is a research-grade leucyl-tRNA synthetase inhibitor used to investigate antibacterial activity against multidrug-resistant gram-negative pathogens. The compound is supplied as a hydrochloride salt with confirmed analytical characterization and is typically provided in small research pack sizes for in vitro studies.
  • Targets leucyl-tRNA synthetase with reported IC50 0.31 μM.
  • High analytical purity (99.6% by LCMS).
  • Supplied as the hydrochloride salt for improved handling and solubility.
  • Intended for antibacterial and LeuRS-target biology research.
  • Chemical formula C11H17BClNO4; molecular weight 273.52 g/mol.
  • Available in small research pack sizes suitable for in vitro assays.
Catalog No. 50-002-39473