Learn More
Bioss EphA2 - Extracellular region M
SDP
Supplier:  Bioss BSM70480M
Encompass_Preferred
EphA2 (Extracellular region) M060 Antibody Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB M060Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-66075