Learn More
Medchemexpress LLC Animal-free IL-2 protein, human (His) | >95.0% | 100UG
SDP
Supplier:  Medchemexpress LLC HYP7037AF100UG
Encompass_Preferred
Recombinant human interleukin-2 (mature sequence A21-T153) is supplied as an animal-free, C-terminal His-tagged protein expressed in E. coli. Supplied lyophilized in a stabilizing buffer, it is >95% pure and intended for research applications requiring animal-free cytokines. Store at -20°C and aliquot for long-term storage.
Catalog No. 50-001-76583