Learn More
Medchemexpress LLC Ephrin-B1/EFNB1 Mou 50ug
SDP
Supplier:  Medchemexpress LLC HYP7302750UG
Encompass_Preferred
Ephrin-B1 (EFNB1) protein binds to Eph family receptors enabling signaling between receptor-expressing cells and cells with EFNB1 It is crucial for neuronal axon growth and expressed in various tissues like the adult lung and colon highlighting its widespread presence and importance in mediating cellular interactions Ephrin-B1/EFNB1 Protein Mouse (HEK293 Fc) is the recombinant mouse-derived Ephrin-B1/EFNB1 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-80778