Learn More
Medchemexpress LLC Epiregulin Human H 5ug
SDP
Supplier:  Medchemexpress LLC HYP730315UG
Encompass_Preferred
Epiregulin Protein a ligand for EGFR and ERBB4 crucially influences inflammation wound healing tissue repair and oocyte maturation It stimulates tyrosine phosphorylation of both receptors regulating angiogenesis vascular remodeling and cell proliferation Interactions with EGFR and ERBB4 contribute to intricate signaling pathways essential for diverse physiological responses and cellular functions Epiregulin Protein Human (HEK293 Fc) is the recombinant human-derived Epiregulin protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-80991