Learn More
Medchemexpress LLC Ephrin Receptor B1 antibody (YA2176) | 10 UL

Supplier: Medchemexpress LLC HYP8243110UL
Ephrin Receptor B1 antibody (YA2176) is intended for laboratory chemicals and the manufacture of substances. This product is for research use only and should only be handled by suitably qualified experienced scientists in appropriately equipped and authorized facilities.
- Host: Rabbit
- Reactivity: Human, mouse, rat
- Clonality number: YA2176
- SwissProt ID: P54762
- Gene ID: 2047
- Appearance: Liquid
- Concentration: 0.3 mg/mL
- Conjugation: Non-conjugated
- Applications: WB, IP
- Dilution ratio: WB: 1:500-1:1000; IP: 1:50
- Formulation: Supplied in rabbit IgG in 10 mM PBS, pH 7.4, 150 mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol.
- Storage and stability: Stored at -20 °C for 1 year; avoid repeated freeze/thaw cycles.
- Shipping: With blue ice
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.