Learn More
Medchemexpress LLC Ephrin Receptor B4 Antibody (YA2892) | 10 UL
SDP
Supplier:  Medchemexpress LLC HYP8314710UL
Encompass_Preferred
Ephrin Receptor B4 Antibody (YA2892) is a Rabbit-derived, non-conjugated IgG monoclonal antibody that targets Ephrin Receptor B4. This antibody is also known by synonyms such as EPHB4, HTK, MYK1, TYRO11, Ephrin type-B receptor 4, Hepatoma transmembrane kinase, and Tyrosine-protein kinase TYRO11.
  • Used in WB and IHC-P applications
  • Reacts with human, mouse, and rat samples
Catalog No. 50-004-45010