Learn More
Medchemexpress LLC EPI-7170 | 2139288-26-7 | 99.9% | 540.88 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1501025MG
Encompass_Preferred
EPI-7170 is a ralaniten analogue and a potent antagonist of the androgen receptor (AR) N-terminal structural domain. It effectively blocks the transcriptional activity of both full-length AR (FL-AR) and AR splice variants (AR-Vs). It has demonstrated antitumor effects, particularly against enzalutamide-resistant castration-resistant prostate cancer (CRPC).
  • Inhibits cell proliferation.
  • Enhances the effect of enzalutamide.
  • Synergistically inhibits AR transcriptional activity.
  • Increases G1 phase and decreases S phase, reducing protein expression levels.
  • Shows anti-tumor activity in vivo.
  • Significantly reduces tumor volume.
Catalog No. 50-003-59316