Learn More
TARGETMOL CHEMICALS INC EPIBERBERINE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T5S236125MG
Encompass_Preferred
Also available in 1 mg 5 mg 10 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. 1. Epiberberine may be caused drug interactions based on CYP2D6 enzyme. 2. Epiberberine has anti-adipogenic effect is mediated by downregulation of the Raf/MEK1/ERK1/2 and AMPK(alpha)/Akt pathways during 3T3-L1 adipocyte differentiation. 3. Epiberberine as the brightest FSA emitter among the alkaloids can also serve as an efficient conformation probe for HTG DNA and discriminate the DNA G-quadruplex from the RNA counterpart. purity: 99%
Catalog No. 50-257-9707