Learn More
Selleck Chemical LLC (-)-Epicatechin gallate 25mg 1257-08-5 ()-ECG, ()-Epicatechin 3-gallate, ()-Epicatechin-3-O-gallate
SDP
Supplier:  Selleck Chemical LLC S392525MG
Encompass_Preferred
(-)-Epicatechin gallate (ECG) is an inhibitor of cyclooxygenase-1 (COX-1) with IC50 of 7.5 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8297