Learn More
Apexbio Technology LLC Epidermal Growth Factor Receptor Peptide (985-996) 96249-43-3 5mg
SDP
Supplier:  Apexbio Technology LLC A10215
Encompass_Preferred
Epidermal Growth Factor Receptor Peptide (985-996) (CAS 96249-43-3) corresponds to a C-terminal segment of the EGFR protein involved in receptor autophosphorylation EGFR a transmembrane receptor tyrosine kinase undergoes activation upon ligand binding triggering receptor dimerization and subsequent phosphorylation of specific tyrosine residues within its C-terminus including the 985-996 region This phosphorylation facilitates recruitment of SH2 domain-containing signaling proteins thereby initiating pathways such as MAPK Akt and JNK which regulate cell proliferation migration and adhesion This peptide serves as a useful research tool for elucidating EGFR-mediated signaling and studying kinase-substrate interactions in cellular models
Catalog No. 50-000-93311