Learn More
Selleck Chemical LLC (-)-epigallocatechin S3922-5mg
SDP
Supplier:  Selleck Chemical LLC S39225MG
Encompass_Preferred
(-)-Epigallocatechin widespread in plants has been shown to exhibit anti-tumor anti-cancer and anti-inflammatory functions
Catalog No. 50-310-9254