Learn More
Apexbio Technology LLC Epigallocatechin 970-74-1 20mg
SDP
Supplier:  Apexbio Technology LLC N187220
Encompass_Preferred
Epigallocatechin (CAS 970-74-1) is a naturally occurring polyphenolic compound isolated from green tea It is known to modulate multiple cellular pathways including oxidative stress response and signal transduction primarily through its antioxidant properties and its ability to influence enzyme activities and cellular signaling cascades Soluble in methanol and ethanol epigallocatechin can be stored under cool dry and airtight conditions This compound is widely utilized in biomedical research for investigating its potential roles in cellular protection cancer biology metabolic regulation and as a tool for studying redox-mediated cellular processes
Catalog No. 50-000-89626