Learn More
Medchemexpress LLC HY-13653 100mg Medchemexpress, (-)-Epigallocatechin Gallate EGCG;Epigallocatechol Gallate CAS:989-51-5 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY13653100MG
Encompass_Preferred
Medchemexpress,HY-13653 100mg (-)-Epigallocatechin Gallate EGCG;Epigallocatechol Gallate CAS:989-51-5 Formula:C22H18O11 (-)-Epigallocatechin Gallate is an antioxidant polyphenol flavonoid form green tea, and inhibits the activation of EGFR, HER2 and HER3, with antitumor activity. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8008