Learn More
Selleck Chemical LLC Epigenetics Compound Library L1900-2X100UL/WELL
SDP
Supplier:  Selleck Chemical LLC L19002X100UL/WELL
Encompass_Preferred
A unique collection of 912 compounds with biological activity used for epigenetic research and associated assays
Catalog No. 50-000-40810
Add to Cart
Request a Quote