Learn More
Medchemexpress LLC Epimedin A | 110623-72-8 | MFCD30496681 | 99.4% | 838.80 | C39H50O20 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN025710MG
Encompass_Preferred
Epimedin A is a natural flavonoid glycoside isolated from Herba Epimedii and provided as a research-grade standard for biochemical and pharmacological studies. It is commonly used in research on osteoclastogenesis, bone resorption, and inflammatory pathways. The product is supplied as a solid powder and should be protected from light during storage.
  • High purity suitable for analytical and pharmacological research.
  • Supplied as a solid powder for convenient handling.
  • Applicable to studies of osteoclastogenesis and anti-inflammatory activity.
  • Stable when stored protected from light under recommended conditions.
  • Provided in small pack sizes for pilot and analytical experiments.
Catalog No. 50-002-96361