Learn More
TARGETMOL CHEMICALS INC Epimedin A 20MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T390620MG
Encompass_Preferred
Also available in 1 mL and bulk. Please contact Fisher for quotes. Epimedin A has anti-osteoporosis activities in vivo. Purity 99.24%
Catalog No. 50-288-0434