Learn More
Cayman Chemical EpInhydrotetracyclInhydro 10mg
SDP
Supplier:  Cayman Chemical 2795410 MG
Encompass_Preferred
A degradation product of tetracycline; active against Pseudomonas, Agrobacterium, Moraxella, Bacillus, and E. coli (MIC50s = 0.75-16 mg/L)
Catalog No. 50-293-7438