Learn More
Cayman Chemical -EpInphrInhydrochlorId 5g
SDP
Supplier:  Cayman Chemical 212455 G
Encompass_Preferred
A natural neurotransmitter that is activates adrenoceptors (Kis = 15, 735, and 3,970 nM for α1A-, β2-, and β1-adrenergic receptors, respectively); may induce either contraction or relaxation of vascular smooth muscle, depending on adrenoceptor subtype expression
Catalog No. 50-298-5176