Learn More
Medchemexpress LLC (-)-Epipodophyllotoxin | 4375-07-9 | 10 MG

Supplier: Medchemexpress LLC HYN765410MG
(-)-Epipodophyllotoxin is an antiproliferative agent against cancer cells isolated from American mayapple *Podophyllum peltatum*. It demonstrates GI50s of 0.36 and 0.24 μM in HeLa cells and MCF-7 cells, respectively, and can inhibit mitotic spindle assembly in vitro.
- Purity: 99.95%
- Molecular weight: 414.41
- Formula: C22H22O8
- Appearance: Solid, white to off-white
- Inhibits mitotic spindle assembly in vitro
- Soluble in DMSO (≥ 50 mg/mL)
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.