Learn More
Abcam Histone E x traction Kit, 100 Tests
Supplier:  Abcam AB113476100TEST
Encompass_Preferred
Histone extraction kit is designed for the quick isolation of total histone extracts from cells and tissue samples within 1 hour assay time, suitable for mammalian and other species. - Cited in over 110 publications - Isolates histones from as few as 10^5 cells or 1 mg of tissue, with optimal results from >10^6 cells or >10 mg of tissue - High yield – up to 0.4 mg of histone protein per 10^7 cells or 100 mg of tissue - Useful in western blot analysis of histone modifications and post-translational modifications - Not for ChIP; use a chromatin extraction kit for ChIP preparation
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0468664