Learn More
Cayman Chemical EpItaLNtrIfluoroACTAte sa 10mg
SDP
Supplier:  Cayman Chemical 3917710 MG
Encompass_Preferred
A synthetic tetrapeptide with anti-aging activities; increases the lifespan of D. melanogaster imagoes from 0.001-5% in the culture medium; delays age-related estrous cycle shutdown and decreases the frequency of bone marrow cell chromosomal aberrations in female mice at 1 μg/animal; decreases spontaneous mammary gland and ovarian tumor development and metastasis in aged female mice at 0.1 UG/animal; stimulates melatonin synthesis in the evening and restores the circadian rhythm of cortisol secretion in senescent M. mulatta monkeys at 10 UG/animal
Catalog No. 50-291-8273